Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TH 10785
GREENER_CHOICE

Catalog No. p-7250153
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

OGG1 activator

TH 10785 is an 8-oxo guanine DNA glycosylase 1 (OGG1) activator. It binds in the active site of OGG1 and increases its recruitment to oxidative DNA damage and its repair.

Specifications

Chemical Name or Material N-Cyclohexyl-2-cyclopropyl-4-quinazolinamine
CAS 1002801-51-5
Quantity 10 mg
Target Glycosylase Activators
Molecular Formula C17H21N3
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.