Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TH 588
Click to view available options
Quantity:
5 mg
25 mg
Description
TH 588 is a potent MTH1 inhibitor (IC50 = 5 nM). Induces oxidative DNA damage and reduces survival in cancer cells. Inhibits growth of breast and colorectal cancer xenografts in mice.
Specifications
Specifications
| Chemical Name or Material | N4-Cyclopropyl-6-(2,3-dichlorophenyl)-2,4-pyrimidinediamine |
| CAS | 1609960-31-7 |
| Assay Percent Range | >98% |
| Quantity | 25 mg |
| Molecular Formula | C13H12Cl2N4 |
| Formula Weight | Observed MW: 295.17 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction