Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Thallium(III) trifluoroacetate, 95%

Manufacturer:  Alfa Aesar™ 8791304

Quantity 2g

 View more versions of this product

Catalog No. AA8791304


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

23586-53-0
543.42
UN1707
Thallium (III) trifluoroacetate
Tl(OOCCF3)3
MFCD00000414
Thallic trifluoroacetate

Specifications

23586-53-0
Tl(OOCCF3)3
MFCD00000414
Thallic trifluoroacetate
213°C (dec.)
Thallium (III) trifluoroacetate
Solid
543.42
UN1707
Hygroscopic
2g
95%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit