Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THAP11 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.72 - $494.86

Specifications

Antigen THAP11
Dilution Western Blot 1:10000, Immunohistochemistry 1:300, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:300
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP149463SS
SDP
View Documents
Novus Biologicals
NBP149463SS
0.025 mL
Each for $181.72
Only null left
Add to Cart
 
NBP149463
SDP
View Documents
Novus Biologicals
NBP149463
0.1 mL
Each for $494.86
Only null left
Add to Cart
 
Description

Description

THAP11 Polyclonal specifically detects THAP11 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

THAP11
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Stem Cell Markers
Whole antisera with 0.1% Sodium Azide
CTG-B43a, CTG-B45d, HRIHFB2206, RONIN, THAP domain containing 11, THAP domain-containing protein 11
THAP11
IgG
Unpurified
Human and mouse. Immunogen sequence has 94% identity to rat and 95% identity to bovine.
Western Blot 1:10000, Immunohistochemistry 1:300, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:300
Polyclonal
Antisera
RUO
Human, Mouse
Q96EK4
57215
Partial recombinant human THAP11 protein expressed in E. coli [Swiss-Prot# Q96EK4]
Primary
Store at -20C. Avoid freeze-thaw cycles.
26 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.