Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THAP11 Antibody, Novus Biologicals™
SDP

Catalog No. NBP149463 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149463 0.1 mL
NBP149463SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP149463 Supplier Novus Biologicals Supplier No. NBP149463
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

THAP11 Polyclonal specifically detects THAP11 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen THAP11
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:10000, Immunohistochemistry 1:300, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:300
Formulation Whole antisera with 0.1% Sodium Azide
Gene Accession No. Q96EK4
Gene Alias CTG-B43a, CTG-B45d, HRIHFB2206, RONIN, THAP domain containing 11, THAP domain-containing protein 11
Gene Symbols THAP11
Host Species Rabbit
Immunogen Partial recombinant human THAP11 protein expressed in E. coli [Swiss-Prot# Q96EK4]
Molecular Weight of Antigen 26 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 57215
Test Specificity Human and mouse. Immunogen sequence has 94% identity to rat and 95% identity to bovine.
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.