Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THAP6 Antibody (OTI1F6), HRP, Novus Biologicals™
SDP

Catalog No. NBP274497H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274497H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274497H Supplier Novus Biologicals Supplier No. NBP274497H

Mouse Monoclonal Antibody

THAP6 Monoclonal specifically detects THAP6 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen THAP6
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1F6
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias MGC30052, THAP domain containing 6, THAP domain-containing protein 6
Gene Symbols THAP6
Host Species Mouse
Immunogen Full length recombinant protein of human THAP6 (NP_653322) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 152815.0
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.