Learn More
Description
Includes
A teacher's guide, student worksheets and casebooks
- Activity involves investigating the theft of the school mascot
- Clues include a note, some strange-looking leaves, a mouth guard and some white fibers left at the scene
- Students will have to figure out whether it was taken by one of the rival schools before the big game
- Investigations include analyzing DNA results, using dichotomous keys to identify selected plants, analyzing handwriting, fingerprints, and bite marks, and a fiber flame test
- A trial is staged after finding the culprit
- For middle school students
- Class size: 30 students
Specifications
Specifications
| Product Type | the Case of the Missing Mascot Lab Activity Kit |
| For Use With (Application) | For middle school students |
| Includes | Teacher's guide, student worksheets and casebooks |
| Class Size | 30 Students |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
