Learn More
Johannes Lieder The Hydra
Shop All Johannes Lieder GmbH & Co. KG Products
Click to view available options
Mounting Type:
Cross and Longitudinal Section
Description
- 5350 The Hydra
- 8 Microscope Slides
- Hydra, extended specimen for general body structure, carefully stained and w.m.
- Hydra with one or more buds w.m.
- Hydra, t.s. through body in different levels showing ectoderm with nematocysts, supporting lamella and entoderm
- Hydra, l.s. through body and tentacles
- Hydra with testis t.s.
- Hydra with ovaries t.s.
- Hydra, isolated cells showing the different cell types, nematocysts
- Hydra with food in digestive cavity, t.s.
Specifications
Specifications
Classification | Zoology |
Material | Elastin Stained Glass |
Mounting Type | Cross and Longitudinal Section |
Type | Microscope Slides |
Slide Type | Prepared Slide |