Learn More
Ansell The Lite One™ Surgical Masks
Catalog No. 19035442 Shop All Ansell Products
Click to view available options
Color:
White
Fastening Type:
Earloops
Ties
This item is not returnable. View return policy
This item is not returnable. View return policy
Features a protective three-layer construction for adequate protection when exposure to blood or bodily fluids is not a risk.
  • Non-sterile
  • Latex-free
  • Pleat-style with ties
TRUSTED_SUSTAINABILITY
Specifications
Product Type Medical/Procedural Face Mask
Color White
Fastening Type Ties
Material Fabric
Sterility Non-sterile
Style Pleated
Construction 3-ply
Filter Efficiency 0.99
Latex Free Yes
Model 48100
Pore Size 0.1 μm (PFE), 3 (BFE)μm
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.