Learn More
Johannes Lieder The Pine (Pinus silvestris)
Shop All Johannes Lieder GmbH & Co. KG Products
Click to view available options
Mounting Type:
Cross and Longitudinal Section
Description
- 6070 The Pine (Pinus sp.)
- 12 Microscope Slides
- Pine, root t.s.
- Pine, older woody stem (twig) t.s., with annular rings and resin ducts
- Pine, wood, three sections: transverse, radial, tangential
- Pine, wood cells, macerated and w.m.
- Pine, stem apex l.s., for meristematic tissue and leaf origin
- Pine, leaves (needles) t.s.
- Pine, male cone with pollen l.s.
- Pine, mature pollen grains, w.m. showing wings
- Pine, young female cone, l.s. shows ovules
- Pine, ovule with growing female gametophyte l.s.
- Pine, ovule with archegonia l.s.
- Pine, mature embryo with endosperm, t.s. with cotyledons
Specifications
Specifications
Classification | Botany |
Material | Elastin Stained Glass |
Mounting Type | Cross and Longitudinal Section |
Type | Microscope Slides |
Slide Type | Prepared Slide |