Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THEM2 Rabbit anti-Human, FITC, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB080881 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB080881 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB080881 Supplier Novus Biologicals Supplier No. NBP297055F

Rabbit Polyclonal Antibody

THEM2 Polyclonal specifically detects THEM2 in Human samples. It is validated for ELISA.

Specifications

Antigen THEM2
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias acyl-CoA thioesterase 13PNAS-27, EC 3.1.2.-, HT012, hypothalamus protein HT012, MGC4961, THEM2, Thioesterase superfamily member 2acyl-coenzyme A thioesterase 13
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant THEM2 (NP_060943.1; Met1-Asn140).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55856
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.