Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THEM4 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB221042 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB221042 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB221042 Supplier Novus Biologicals Supplier No. NBP177360V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

THEM4 Polyclonal antibody specifically detects THEM4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen THEM4
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C-terminal modulator protein, CTMPCarboxyl-terminal modulator protein, MGC29636, thioesterase superfamily member 4
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the center of human THEM4. The immunogen is located within amino acids 60 - 110 of THEM4. Amino Acid Squence: EGDSWKRLPSYKRTPTE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 117145
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.