Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Themis Antibody (701010), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB375469 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375469 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375469 Supplier Novus Biologicals Supplier No. FAB6929A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Themis Monoclonal antibody specifically detects Themis in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen Themis
Applications Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 701010
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias bA325O24.3, bA325O24.4, C6orf190, C6orf207, FLJ40584, GASP, GRB2-associated protein, MGC163388, protein THEMIS, signaling phosphoprotein specific for T cells, thymocyte selection associated, thymocyte selection pathway associated, thymocyte-expressed molecule involved in selection, TSEPA
Host Species Mouse
Immunogen E. coli-derived recombinant human Themis, Ala2-Pro282, Accession # Q8N1K5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387357
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.