Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THEMIS Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody

$167.95

Specifications

Antigen THEMIS
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-157-0003
Encompass_Preferred
View Documents
Bethyl Laboratories, Inc
A305277AT
20 μL
Each of 1 for $167.95
Only null left
Add to Cart
 
Description

Description

The recommended shelf life for this product is 1 year from date of receipt.

Themis is a cytoplasmic protein that is essential for positive selection and the development of single-positive thymocytes. Highest levels of expression are found in CD4+CD8+ double-positive thymocytes, with lower expression found in mature T cells. In addition to Grb1, Themis has been found to associate with PLC gamma 1 and LAT and is quickly phosphorylated by SLP-76 following TCR signaling. There are 5 isoforms of mouse Themis that are produced as a result of alternative splicing.
Specifications

Specifications

THEMIS
Polyclonal
Liquid
RUO
TBS with 0.1% BSA and 0.09% sodium azide
THEMIS; thymocyte selection associated; C6orf190; C6orf207; FLJ40584; MGC163388; TSEPA; bA325O24.3; bA325O24.4; thymocyte selection pathway associated; thymocyte-expressed molecule involved in selection
THEMIS
IgG
Antigen affinity chromatography
Antibody
Western Blot
Unconjugated
Rabbit
Human
Q8N1K5
387357
Between 591 and 641
Primary
4° C
THEMIS
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.