Learn More
Therapak™ Aqui-Pak™ Absorbent Sleeves
Shop All Therapak Corporation Products
                    Click to view available options
                                Quantity:
                                Up to four tubes
                                Up to six tubes
                    Description
Ideal for Biological Substance and Exempt Human specimen shipments
- For safe transport of specimen tubes in accordance with DOT/IATA regulations
 - Consist of multilayered polymeric material with extremely high absorption properties
 - Simultaneously separates and cushions specimen tubes
 - Place between primary and secondary container
 - Roll it up and place it in a bag or canister
Specifications
Specifications
| Quantity | Up to six tubes | 
| Type | Absorbent Sleeve | 
| Capacity | 275 mL | 