Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Therapak™ Biological Substance Category B Replenishment Supplies

Catalog No. 22130058 Shop All Therapak Corporation Products
Click to view available options
Includes:
3 oz. Refrigerant Pack
8 oz. Refrigerant Pack
Large Insulated EPS Cooler
Medium Insulated EPS Cooler
Small Insulated EPS Cooler
Size:
3 oz. pack
8 oz. pack
Large
Medium
Small

For shipment of biological specimens

  • Insulated Shippers include standard DOT/IATA labels, EPS cooler, and corrugated shipper box
  • Leakproof Refrigerant Shippers keep lab specimens cold (30°F) with nontoxic, food-grade gel refrigerants

Specifications

Includes Small Insulated EPS Cooler
Applications For shipment of Biological Substance Category B specimens
Size Small
Dimensions (LxWxH) Interior: 8 x 6 x 8 in.Exterior: 11 x 9 x 11 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.