Learn More
Therapak™ Single Specimen Collection Kits
Catalog No. 22131555 Shop All Therapak Corporation Products
Click to view available options
Includes:
Evidence Seal
Temp. Strip, Evidence Seal, Bag
Temperature Strip
Temperature Strip, Evidence Seal
Temperature Strip, Transport Bag
This item is not returnable. View return policy
This item is not returnable. View return policy
Single Specimen Collection Kits can be used in conjunction with on-site testing products or remote site collection when specimen is transported to a laboratory for analysis.
Single Specimen Collection Kits can be used in conjunction with on-site testing products or remote site collection when specimen is transported to a laboratory for analysis. Each kit contains either a 180 mL beaker style collection cup with pour spout or a leak-resistant 90 mL sterile container with cap and integrity seal. The transport bag features two sealable, tamper evident pockets and comes with absorbent material. The individually bagged kits are available in various configurations.
Specifications
Product Type Urine Specimen Container
Sterility Sterile
For Use With (Application) For drugs of abuse testing, on-site testing or remote site collection
Includes Temperature Strip, Transport Bag
Volume (English) 3 oz.
Volume (Metric) 90 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.