Learn More
Thermo Scientific™ ABsolute™ MAX QRT-PCR Mixes
Provide a quick, reliable, and efficient method for detecting and quantifying RNA
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
AB-1340B
|
Thermo Scientific™
AB1340B |
QRT-PCR ROX mix; 1.2mL vials; 800 reactions | QRT-PCR ROX mix; 1.2mL vials; 800 reactions | N/A | N/A | N/A | |||
AB-1349A
|
Thermo Scientific™
AB1349A |
Capillary dUTP Mix for the Lightcycler; 2 x 1.25mL vials; 250 reactions | Capillary dUTP Mix for the Lightcycler; 2 x 1.25mL vials; 250 reactions | N/A | N/A | N/A | |||
AB-1349B
|
Thermo Scientific™
AB1349B |
Capillary dUTP Mix for the Lightcycler; 16 x 1.25mL vials; 2000 reactions | Capillary dUTP Mix for the Lightcycler; 16 x 1.25mL vials; 2000 reactions | N/A | N/A | N/A | |||
Description
- Formulated for use in one-step QRT-PCR reactions, with gene-specific primers and probes
- Minimal pipetting required—protocol is easy to follow and automation is viable
- 2X mix is supplied with a proprietary blend of RTases combining the advantages of AMV and MMuLV RTases to give unparalleled performance
- Can be used at elevated temperatures, avoiding problems caused by secondary structure in the RNA template
- QRTase Blend has greater dynamic range than either AMV or MMuLV, leading to greater accuracy in QRT-PCR experiments
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.