Learn More
Thermo Scientific™ ABsolute™ QPCR Low ROX™ Mixes
Optimized for use with real-time instruments that are able to detect long wavelength dyes with greater sensitivity (ABI™ 7500)
Supplier:  Thermo Scientific™ AB1320B
 View more versions of this product
Catalog No. AB-1320-B
Description
Description
  • Incorporate Thermo-Start™ DNA polymerase, a hot-start enzyme that provides superior specificity and excellent sensitivity
  • Utilize a proprietary reaction buffer that provides high sensitivity, specificity and reproducibility
  • Contain all the components necessary to perform quantitative PCR, with the exception of template and primers/probes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
800
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.