Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ ABsolute™ QPCR Low ROX™ Mixes
Optimized for use with real-time instruments that are able to detect long wavelength dyes with greater sensitivity (ABI™ 7500)
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
AB-1320-A
|
Thermo Scientific™
AB1320A |
dUTP, 100 reactions; 2 x 1.25mL | dUTP, 100 reactions; 2 x 1.25mL | N/A | N/A | N/A | |||
AB-1320-B
|
Thermo Scientific™
AB1320B |
dUTP, 800 reactions; 16 x 1.25mL | dUTP, 800 reactions; 16 x 1.25mL | N/A | N/A | N/A | |||
AB-1321-A
|
Thermo Scientific™
AB1321A |
dUTP, 200 reactions; 1 x 5mL | dUTP, 200 reactions; 1 x 5mL | N/A | N/A | N/A | |||
AB-1321-B
|
Thermo Scientific™
AB1321B |
dUTP, 2000 reactions; 10 x 5mL | dUTP, 2000 reactions; 10 x 5mL | N/A | N/A | N/A | |||
AB-1324-A
|
Thermo Scientific™
AB1324A |
dUTP, 100 reactions; 2 x 1.25mL | dUTP, 100 reactions; 2 x 1.25mL | N/A | N/A | N/A | |||
AB-1324-B
|
Thermo Scientific™
AB1324B |
dUTP, 800 reactions; 16 x 1.25mL | dUTP, 800 reactions; 16 x 1.25mL | N/A | N/A | N/A | |||
AB-1325-A
|
Thermo Scientific™
AB1325A |
dUTP, 200 reactions; 1 x 5mL | dUTP, 200 reactions; 1 x 5mL | N/A | N/A | N/A | |||
AB-1325-B
|
Thermo Scientific™
AB1325B |
dUTP, 800 reactions; 16 x 1.25mL | dUTP, 800 reactions; 16 x 1.25mL | N/A | N/A | N/A | |||
Description
- Incorporate Thermo-Start™ DNA polymerase, a hot-start enzyme that provides superior specificity and excellent sensitivity
- Utilize a proprietary reaction buffer that provides high sensitivity, specificity and reproducibility
- Contain all the components necessary to perform quantitative PCR, with the exception of template and primers/probes
- Practice of the patented 5apostrophe Nuclease Process requires a license from Applied Biosystems. The purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaserapostrophes own internal research when used with the separate purchase of Licensed Probe. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained from the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
- Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaserapostrophes own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title