Learn More
Thermo Scientific™ Aquasil™ C18 Reversed Phase HPLC Column, 5 μm, 2.1 mm x 50 mm
Description
Thermo Scientific HPLC columns are manufactured under strict ISO 9001:2000 procedures, and every batch of silica and bonded phase is thoroughly tested for physical and chromatographic characteristics. Test reports are included with every column shipped.
- The combination of a moderate coverage C18 group and an additional polar group gives new selectivity compared to a traditional C18 phase
- Can also be used with fewer additives in the mobile phase, perhaps even no additives at all, depending on application
- Compatible with 100% aqueous mobile phases
- Excellent peak shapes for basic, acidic and neutral compounds
- Excellent results with low buffer concentrations
- Stable for LC/MS applications
Specifications
Specifications
| Column Format | Javelin Express Column |
| Diameter (Metric) | 2.1 mm |
| Length (Metric) | 50 mm |
| Particle Size | 5 μm |
| Pore Size | 100 Å |
| USP Type | L1 |
| Stationary Phase | C18 |
| Packing Material | Silica |
| Carbon Load | 12% |
| Column Type | Reversed Phase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.