Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BetaBasic™ 18 DASH-HTS HPLC Columns, 5μm Particle Size

Catalog No. 71505012150
Change view
Click to view available options
Particle Size: 5μm; 10L x 2.1mm I.D.; 1/pk.
Particle Size: 5μm; 20L x 2.1mm I.D.; 1/pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
71505012150 Particle Size: 5μm; 10L x 2.1mm I.D.; 1/pk.
71505022150 Particle Size: 5μm; 20L x 2.1mm I.D.; 1/pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71505012150 Supplier Thermo Scientific™ Supplier No. 71505012150

Designed for high throughput, these short columns are excellent for ballistic gradients, with retention and sensitivity for very fast LC/MS applications

  • Patented, low dead volume design minimizes band spreading
  • Finger-tight fittings make changing columns quick and easy

Features of BetaBasic 18 Solid Phase

  • Highly efficient and reproducible columns
  • Pore size suitable for small molecules, peptides and protein digests
  • Superb pH stability

TRUSTED_SUSTAINABILITY

Specifications

Column Format DASH-HTS Column, 1/Pk.
Diameter (Metric) 2.1 mm
Length (Metric) 10 mm
Particle Size 5 μm
pH 2 to 12
Pore Size 150 Å
Surface Area 200 m²/g
USP Type L1
Stationary Phase C18
Packing Material Silica
Carbon Load 13%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.