Learn More
Thermo Scientific™ BetaBasic™ C18 Reversed Phase HPLC Column, 3 μm, 10 mm x 150 mm
Description
- Pore size suitable for small molecules, peptides and protein digests
- Superb pH stability
Order Info
Other column dimensions are also available. Please call Customer Service for more information.
Specifications
Specifications
| Diameter (Metric) | 10 mm |
| Length (Metric) | 150 mm |
| Particle Size | 3 μm |
| pH | 2 to 12 |
| Pore Size | 150 Å |
| Surface Area | 200 m²/g |
| USP Type | L1 |
| Stationary Phase | C18 |
| Packing Material | Silica |
| Carbon Load | 13% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.