Learn More
Thermo Scientific™ BetaBasic™ 18 C18 Reversed Phase KAPPA HPLC Column, 5 μm, 0.32 mm x 100 mm
Description
- Rugged sleeved design for robustness
- Excellent for LC/MS analysis
Features of BetaBasic 18 Solid Phase
- Highly efficient and reproducible columns
- Pore size suitable for small molecules, peptides and protein digests
- Superb pH stability
Specifications
Specifications
| Column Format | Capillary Column |
| Diameter (Metric) | 0.32 mm |
| Length (Metric) | 100 mm |
| Particle Size | 5 μm |
| pH | 2 to 12 |
| Pore Size | 150 Å |
| Surface Area | 200 m²/g |
| USP Type | L1 |
| Stationary Phase | C18 |
| Packing Material | Silica |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.