Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BetaBasic 4 CPF HPLC Columns

Catalog No. 71605151032
Change view
Click to view available options
Particle Size: 5μm; 150 x 1.0mm I.D.
Particle Size: 5μm; 150 x 2.1mm I.D.
Particle Size: 5μm; 150 x 3.0mm I.D.
Particle Size: 5μm; 150 x 4.6mm I.D.
Particle Size: 5μm; 250 x 1.0mm I.D.
Particle Size: 5μm; 250 x 2.1mm I.D.
Particle Size: 5μm; 250 x 3.0mm I.D.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
71605151032 Particle Size: 5μm; 150 x 1.0mm I.D.
71605152132 Particle Size: 5μm; 150 x 2.1mm I.D.
71605153032 Particle Size: 5μm; 150 x 3.0mm I.D.
71605154632 Particle Size: 5μm; 150 x 4.6mm I.D.
71605251032 Particle Size: 5μm; 250 x 1.0mm I.D.
71605252132 Particle Size: 5μm; 250 x 2.1mm I.D.
71605253032 Particle Size: 5μm; 250 x 3.0mm I.D.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 71605151032 Supplier Thermo Scientific™ Supplier No. 71605151032

A good first choice for compounds with poor peak symmetry on other columns

Thermo Scientific HPLC columns are manufactured under strict ISO 9001:2000 procedures, and every batch of silica and bonded phase is thoroughly tested for physical and chromatographic characteristics. Test reports are included with every column shipped.

  • With Integral Column Plus Filter (CPF) System
  • Fully deactivated Type B silica to ensure sharp peaks and excellent reproducibility
  • Give less retention than C18 or C8 phases, and are a good choice for fast analyses including LC/MS of all types of compounds
  • For basic compounds at high pH, which allows for a wide choice of buffer selection in LC/MS applications
  • Stable bonding chemistry and excellent surface coverage gives the columns exceptional low and high pH stability

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 1 mm
Length (Metric) 150 mm
Particle Size 5 μm
Pore Size 150 Å
USP Type L26
Stationary Phase C4
Packing Material Silica
Carbon Load 6%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.