Learn More
Thermo Scientific™ BetaBasic 4 CPF HPLC Columns
Description
Thermo Scientific HPLC columns are manufactured under strict ISO 9001:2000 procedures, and every batch of silica and bonded phase is thoroughly tested for physical and chromatographic characteristics. Test reports are included with every column shipped.
- With Integral Column Plus Filter (CPF) System
- Fully deactivated Type B silica to ensure sharp peaks and excellent reproducibility
- Give less retention than C18 or C8 phases, and are a good choice for fast analyses including LC/MS of all types of compounds
- For basic compounds at high pH, which allows for a wide choice of buffer selection in LC/MS applications
- Stable bonding chemistry and excellent surface coverage gives the columns exceptional low and high pH stability
Specifications
Specifications
| Diameter (Metric) | 3 mm |
| Length (Metric) | 150 mm |
| Particle Size | 5 μm |
| Pore Size | 150 Å |
| USP Type | L26 |
| Stationary Phase | C4 |
| Packing Material | Silica |
| Carbon Load | 6% |
| Column Type | Reversed Phase |
| Endcapped | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.