Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BetaBasic™ 4 HPLC Columns, 3μm Particle Size

Catalog No. 71603053030
Change view
Click to view available options
Length (Metric):
50 mm
150 mm
Diameter (Metric):
1 mm
3 mm
4.6 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
71603053030 50 mm 3 mm
71603051030 50 mm 1 mm
71603154630 150 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 71603053030 Supplier Thermo Scientific™ Supplier No. 71603053030

A good first choice for compounds with poor peak symmetry on other columns

  • 150Å pore size for small molecules, peptides and protein digests
  • Superb pH stability

Order Info

Other column dimensions are also available. Please call Customer Service for more information.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 3 mm
Length (Metric) 50 mm
Particle Size 3 μm
Pore Size 150 Å
USP Type L26
Stationary Phase C4
Packing Material Silica
Carbon Load 6%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.