Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ BetaBasic™ 8 Guard Cartridges
Click to view available options
Diameter (Metric) Inner:
1 mm
3 mm
Description
- The 10mm design offers maximum protection with minimal increase in retention
- Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
- Highly efficient and reproducible Thermo Scientific columns
- 150Å pore size for small molecules, peptides and protein digests
- Superb pH stability

Specifications
Specifications
Particle Size | 5 μm |
Length (Metric) | 10 mm |
Diameter (Metric) Inner | 1 mm |
Stationary Phase | BetaBasic 8 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction