Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BetaBasic™ CN CPF HPLC Columns

Catalog No. 71705152132
Change view
Click to view available options
Particle Size: 5μm; 150 x 1.0mm I.D.
Particle Size: 5μm; 150 x 2.1mm I.D.
Particle Size: 5μm; 150 x 3.0mm I.D.
Particle Size: 5μm; 150 x 4.6mm I.D.
Particle Size: 5μm; 250 x 1.0mm I.D.
Particle Size: 5μm; 250 x 2.1mm I.D.
Particle Size: 5μm; 250 x 3.0mm I.D.
Particle Size: 5μm; 250 x 4.6mm I.D.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
71705152132 Particle Size: 5μm; 150 x 2.1mm I.D.
71705151032 Particle Size: 5μm; 150 x 1.0mm I.D.
71705153032 Particle Size: 5μm; 150 x 3.0mm I.D.
71705154632 Particle Size: 5μm; 150 x 4.6mm I.D.
71705251032 Particle Size: 5μm; 250 x 1.0mm I.D.
71705252132 Particle Size: 5μm; 250 x 2.1mm I.D.
71705253032 Particle Size: 5μm; 250 x 3.0mm I.D.
71705254632 Particle Size: 5μm; 250 x 4.6mm I.D.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 71705152132 Supplier Thermo Scientific™ Supplier No. 71705152132

A good first choice for compounds typically considered difficult to analyze

Thermo Scientific HPLC columns are manufactured under strict ISO 9001:2000 procedures, and every batch of silica and bonded phase is thoroughly tested for physical and chromatographic characteristics. Test reports are included with every column shipped.

  • For small molecules, peptides and protein digests
  • Low pH stability compared to other cyano phases
  • Offer alternative selectivity based on dipole-dipole and dipole-induced dipole interactions, and can be used in normal or reversed phase separations

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 150 mm
Particle Size 5 μm
pH 2 to 9
Pore Size 150 Å
Surface Area 200 m²/g
USP Type L10
Stationary Phase Cyano (CN)
Packing Material Silica
Carbon Load 5%
Column Type Mixed Mode
Endcapped Yes
Phase Normal and Reversed Phase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.