Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BetaBasic™ CN SFC Columns

Catalog No. 71705252160
Change view
Click to view available options
Particle Size: 5μm; 100L x 1.0mm I.D.
Particle Size: 5μm; 100L x 2.1mm I.D.
Particle Size: 5μm; 100L x 4.6mm I.D.
Particle Size: 5μm; 150L x 1.0mm I.D.
Particle Size: 5μm; 150L x 2.1mm I.D.
Particle Size: 5μm; 150L x 4.6mm I.D.
Particle Size: 5μm; 250L x 1.0mm I.D.
Particle Size: 5μm; 250L x 2.1mm I.D.
Particle Size: 5μm; 250L x 4.6mm I.D.
Particle Size: 5μm; 50L x 2.1mm I.D.
Particle Size: 5μm; 50L x 4.6mm I.D.
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
71705252160 Particle Size: 5μm; 250L x 2.1mm I.D.
71705052160 Particle Size: 5μm; 50L x 2.1mm I.D.
71705054660 Particle Size: 5μm; 50L x 4.6mm I.D.
71705101060 Particle Size: 5μm; 100L x 1.0mm I.D.
71705102160 Particle Size: 5μm; 100L x 2.1mm I.D.
71705104660 Particle Size: 5μm; 100L x 4.6mm I.D.
71705151060 Particle Size: 5μm; 150L x 1.0mm I.D.
71705152160 Particle Size: 5μm; 150L x 2.1mm I.D.
71705154660 Particle Size: 5μm; 150L x 4.6mm I.D.
71705251060 Particle Size: 5μm; 250L x 1.0mm I.D.
71705254660 Particle Size: 5μm; 250L x 4.6mm I.D.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 71705252160 Supplier Thermo Scientific™ Supplier No. 71705252160

Highly efficient and reproducible

  • Pore size suitable for small molecules, peptides and protein digests
  • Superb pH stability

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 250 mm
Particle Size 5 μm
pH 2 to 9
Pore Size 150 Å
Surface Area 200 m²/g
USP Type L10
Stationary Phase Cyano (CN)
Packing Material Silica
Carbon Load 5%
Column Type Mixed Mode
Endcapped Yes
Phase Reversed Phase, Normal Phase, Supercritical Fluid Chromatography
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.