Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Easy Pierce Heat Sealing Foil

Catalog No. AB1797
Click to view available options
Material:
Aluminum
Polyester
Product Type:
Clear Seal Thin
Easy Peel Film Laminate
Easy Pierce Foil
EasyPierce 20μm Thin Foil
Dimensions (L x W):
80 x 125 mm
85 x 135 mm

Use this heat sealing foil to ready your microplate for compound management and high-throughput screening applications, such as PCR.

Seal your microplate for compound management and high-throughput screening, including PCR, with Thermo Scientific™ Easy Pierce Heat Sealing Foil. Suitable for use with Dimethyl sulfoxide (DMSO) reagents, this sealing foil creates a peelable bond that can be easily pierced and removed by hand.

Features:

  • Quick operation
  • Safe for temperature-sensitive samples
  • Designed for reliable performance with Thermo Scientific ALPS instruments, including the, ALPS 3000, ALPS 25 and ALPS 50 V
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Solvent Or Alcohol Resistant Life Science Reagent Resistant
Material Polyester
Sterility Non-sterile
Autoclavable Not Autoclavable
No. of Wells 96
No. per Case 1000
No. per Pack 1000
Dimensions (L x W) 80 x 125 mm
For Use With (Application) Ideal for fluorescence, colorimetric, and optical applications; suitable for PCR/ qPCR; ELISA
Content And Storage General; Long term
For Use With (Equipment) ALPS 3000, ALPS 25 and ALPS 50 V; AB-1443A
Product Type Clear Seal Thin
Product Line Abgene
Adhesive Polypropylene
Certified RNase/DNase-free No
Pierceable Yes
Temperature Range (Metric) -80°C to +120°C
Well Shape All well shapes
Plate Compatibility Polypropylene, Polyethylene, Polystyrene
Incubation Conditions General
Peelable Yes
Fluorescence Low Autofluorescence
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.