Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phire Tissue Direct PCR Master Mix

Catalog No. p-4861495
Change view
Click to view available options
No. of Reactions:
1250 Reactions
250 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
F170S 250 Reactions
F170L 1250 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F170S Supplier Thermo Scientific™ Supplier No. F170S
Only null left
Add to Cart
Edge
Add to Cart

Thermo Scientific Phire Tissue Direct PCR Master Mix has been developed for amplification of DNA directly from a wide variety of tissues obtained from mice, human, fish, birds and insects.

Thermo Scientific Phire Tissue Direct PCR Master Mix has been developed for amplification of DNA directly from a wide variety of tissues obtained from mice, humans, fish, birds, and insects. The Master Mix containing Phire Hot Start II DNA Polymerase is specially formulated to perform PCR in the presence of different animal tissue–derived inhibitors such as collagen, melanin and eumelanin (hair, skin), and myoglobin (muscle). The kit also includes DNARelease Additive, which can be used to improve the release of DNA from difficult tissues.

Features

  • Direct PCR reduces time-consuming and expensive DNA purification steps
  • Available Dilution & Storage protocol allows multiple PCR reactions from a single sample
  • Specially engineered Phire DNA polymerase for extremely short PCR protocol times
  • Master mix format with premixed gel loading dye minimizes cross-contamination, reduces sample handling, and allows direct loading on gel

Applications

  • Genotyping
  • Transgene detection
  • Knockout analysis
  • Sequencing

Notes

  • The optimal annealing temperature for Phire DNA Polymerases may differ significantly from that of Taq-based polymerases.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • 2X Phire Tissue Direct PCR Master Mix, 2 x 1.25 mL
• Dilution Buffer, 5 mL
• DNARelease Additive, 3 x 100 μL
• 25 μM Control Primer Mix, 40 μL
• Nuclease-free water, 2 x 1.25 mL
• 50 μg O'GeneRuler Express DNA Ladder

Store at -20°C.
GC-Rich PCR Performance Medium
PCR Method Direct PCR
Polymerase Phire Hot Start II DNA Polymerase
Reaction Speed Fast
Includes Label or Dye Yes
Product Type Direct PCR Master Mix
Quantity 250 reactions
Shipping Condition Wet or Dry Ice
For Use With (Application) Genotyping, Hot-start PCR
Hot Start Built-In Hot Start
No. of Reactions 250 Reactions
Overhang Blunt
Reaction Format SuperMix or Master Mix
Sample Type Tissue
Starting Material DNA
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.