Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phusion U Hot Start DNA Polymerases

Catalog No. F556L
Click to view available options
Color:
Colorless
Green
No. of Reactions:
100 Reactions
500 Reactions

Catalog No. F556L

Supplier: Thermo Scientific™ F556L

Only null left
Add to Cart
Add to Cart

Thermo Scientific Phusion U DNA polymerase is a novel engineered high fidelity enzyme developed using fusion technology.

Thermo Scientific Phusion U DNA Polymerase is a novel engineered high fidelity enzyme developed using fusion technology. Due to a proprietary mutation in the so called dUTP binding pocket of Phusion polymerase, Phusion U polymerase overcomes an important limitation of proofreading enzymes: it is able to incorporate dUTP and read through uracil present in DNA templates.

In addition to the uracil usage possibility, Phusion U polymerase features all the superior properties of other Phusion DNA polymerases: great accuracy, speed, ability to amplify long amplicons up to 20 kb, and a high specificity with Affibody-based hot start. These features make Phusion U Hot Start DNA Polymerase an excellent choice for such important applications as amplification of bisulphite-converted or damaged DNA as well as use of carryover contamination control.

Phusion U Hot Start Green DNA Polymerase is a combination of Phusion U Hot Start DNA Polymerase and 5X Phusion Green buffers. The buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with Phusion U polymerase performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

Highlights

  • Accuracy—high fidelity DNA polymerase (25X Taq)
  • Uracil-tolerance—engineered to incorporate dUTP and amplify uracil containing templates
  • Specificity—hot start for reduced nonspecific amplification and primer degradation
  • Speed—short extension times (15–30 s/kb)
  • Green format—permits direct loading of PCR products on gels

Applications

  • Amplification of bisulphite-converted DNA
  • Amplification of damaged or aged DNA
  • Carry-over contamination control
  • Uracil-excision based (USER) cloning methods

Using Phusion DNA polymerases

Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 U/μL
Content And Storage • Phusion U Hot Start DNA Polymerase, 2 U/μL
• 5X Phusion Green HF & GC Buffers
• DMSO
• 50 mM MgCl2 solution

Phusion HF Buffer and Phusion GC Buffers each provide 1.5 mM MgCl2 in the final 1X concentration.
Store at -20°C.
GC-Rich PCR Performance High
Polymerase Phusion U Hot Start DNA Polymerase
Reaction Speed Fast
Product Type Hot Start DNA Polymerase
Quantity 500 units
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq) 25X
Hot Start Built-In Hot Start
No. of Reactions 500 Reactions
Overhang Blunt
Reaction Format Standalone
Size (Final Product) 20 kb or less
Color Green
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.