Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phusion U Hot Start PCR Master Mix

Catalog No. F533L
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions

Thermo Scientific Phusion U DNA polymerase is a novel engineered high fidelity enzyme developed using fusion technology.

Thermo Scientific Phusion U Hot Start PCR Master Mix is a convenient 2X formula that contains Phusion U Hot Start DNA Polymerase, nucleotides, and an optimized reaction buffer including MgCl2.

Phusion U DNA Polymerase is a novel engineered high-fidelity enzyme developed using fusion technology. Designed with a proprietary mutation in the dUTP binding pocket of Phusion polymerase, Phusion U polymerase is able to overcome important limitations of proofreading enzymes to incorporate dUTP and read through uracil present in DNA templates.

Phusion U polymerase features properties of Phusion DNA polymerases and offers the ability to amplify long amplicons up to 20 kb and a high specificity with affibody based hot start.

These features make Phusion U Hot Start DNA Polymerase an excellent choice for such applications as amplification of bisulfite-converted or damaged DNA as well as use of carryover contamination control.

Highlights

  • High-fidelity DNA polymerase (25X Taq)
  • Engineered to incorporate dUTP and amplify uracil containing templates
  • Hot start for reduced nonspecific amplification and primer degradation
  • Only template and primers need to be added for use
  • Short extension times (15–30 s/kb)

Applications

  • Amplification of bisulfite-converted DNA
  • Amplification of damaged or aged DNA
  • Carry-over contamination control
  • Uracil-excision based (USER) cloning methods

Using Phusion DNA Polymerases

  • Annealing rules for Phusion DNA polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • Phusion U Hot Start DNA Polymerase
• 2X reaction buffer
• 3 mM MgCl2
• 400 μM each of dATP, dTTP, dCTP and dGTP

Master mix is supplied with nuclease-free water.
Store at -20°C.
GC-Rich PCR Performance Medium
Polymerase Phusion U Hot Start DNA Polymerase
Reaction Speed Fast
Product Type Hot Start PCR Master Mix
Quantity 500 reactions
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR
Fidelity (vs. Taq) 25X
Hot Start Built-In Hot Start
No. of Reactions 500 Reactions
Overhang Blunt
Reaction Format SuperMix or Master Mix
Size (Final Product) 20 kb or less
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.