Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Hypersil BDS CN CPG HPLC Columns--5μm Particle Size
Supplier: Thermo Scientific™ 28805104631
Specifications
4.6 mm | |
5 μm | |
Cyano (CN) | |
Mixed Mode |
100 mm | |
L10 | |
Base-Deactivated Silica | |
Normal and Reversed Phase |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title