Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Membrane Endo Agar LES (Dehydrated)

Catalog No. oxmm0551b
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXMM0551B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXMM0551B Supplier Thermo Scientific™ Supplier No. MM0551B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Enumerate coliform organisms in water samples using this medium.

Enumerate coliform organisms in water samples using Thermo Scientific™ Oxoid™ Membrane Endo Agar LES (Dehydrated) when used with basic Fuschin (BR0050). This membrane filtration medium is prepared according to the Lawrence Experimental Station formulation of McCarthy, Delaney and Grasso1 and used for the enumeration of coliform organisms in water2 using a 2-stage enrichment technique.

Use Membrane Endo Agar LES for enumeration of coliform organisms in water.

The value of the membrane filter technique for the enumeration of coliform organisms in water lies in its greater reliability and precision when compared with the MPN multiple tube test3.
McCarthy, Delaney and Grasso1 have recommended a two-stage process of enrichment to provide a non-toxic environment for maximal resuscitation of the coliforms.

Calabrese and Bissonnette4 found that supplementation of M-Endo medium with catalase and sodium pyruvate resulted in improved recovery of coliform bacteria from chlorinated sewage effluent.

Experiments carried out by Noble5 indicated that sodium sulphite and Basic Fuchsin can be extremely detrimental to the stressed coliforms, reducing the total count.

Enrichment for a period of 2 hours ± 0.5 hours in single strength Lauryl Tryptose Broth CM0451 will give adequate resuscitation to the stressed coliform organisms and provide the best assessment of the quality of the drinking water.

Enrichment is usually not necessary for the examination of non-potable waters and sewage effluents.

Selection of the sample volume is governed by the expected bacterial density. An ideal quantity will result in growth of more than 50 coliform colonies and less than 200 colonies of all types.

All organisms which produce a colony with a golden-green metallic sheen within 24 hours incubation are considered members of the coliform group. The sheen may cover the entire colony or be restricted to the central area or the periphery. The recommended depth of M-Endo Agar LES in plates restricts the colony size and hence facilitates carrying out the colony count.

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: APHA standards.
Description Membrane Endo Agar LES
Form Powder
Product Type Agar
Quantity 500 g
Yield For 11.1L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.