Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DSS (disuccinimidyl suberate), No-Weigh™ Format

Catalog No. pi21555
Click to view available options
Quantity:
50 mg
10 x 2 mg
1 g

Crosslink amines to amines with disuccinimidyl suberate composed of NHS-ester groups with a membrane-permeable, mid-length spacer for intracellular protein crosslinking.

Thermo Scientific™ Pierce DSS, also called disuccinimidyl suberate, is noncleavable and membrane permeable crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester at each end of an 8-carbon spacer arm.

Highlights:

  • Chemical crosslinking of intracellular proteins prior to cell lysis and immunoprecipitation
  • “Fix” protein interactions to allow identification of weak or transient protein interactions
  • Protein crosslinking to create bioconjugates via single-step reactions
  • Crosslink oriented antibodies to Protein A or G agarose
  • Immobilize proteins onto amine-coated surfaces
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Amine
Cleavable No
Description DSS
Molecular Weight (g/mol) 368.35
PEGylated No
Spacer Arm Length 11.4 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety NHS Ester
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 1 g
Solubility DMF, DMSO
Format Standard, Single-use
Water Soluble No
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.