Learn More
Thermo Scientific™ Valve with SMART Indicating Purifier (VIP)
Provide either flow control or positive shut off while maintaining gas purity for gasses 99.9999% pure or better
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
60-180-950
|
Thermo Scientific™
60180950 |
Multi-turn Valve | N/A | N/A | N/A | ||||
60-180-952
|
Thermo Scientific™
60180952 |
Quarter Turn Valve with Triple Filter | N/A | N/A | N/A | ||||
60-180-953
|
Thermo Scientific™
60180953 |
Quarter Turn Valve with Combi Filter | N/A | N/A | N/A | ||||
60-180-951
|
Thermo Scientific™
60180951 |
Multi-turn Valve with Combi Filter | N/A | N/A | N/A | ||||
60-180-941
|
Thermo Scientific™
60180941 |
Replacement Combi Cartridge | N/A | N/A | N/A | ||||
Description
- Provides a visual indication of contaminants in the gas stream while it is removing them, ensuring consistent performance of analytical processes
- Safeguards the gas stream and process
- Specially cleaned for all analytical applications and will not generate contaminates that consume the purifier
- Patented Quick-Change Base to allow for easy change out of spent purifiers
- Available in both a multiturn and quarter turn models
- All regulators supplied with 1/8 in. stainless steel output adaptor