Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THG1L Monoclonal Antibody (4F11)
GREENER_CHOICE

Catalog No. PIMA524482
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524482 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524482 Supplier Invitrogen™ Supplier No. MA524482
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MAKSKFEYVR DFEADDTCLA HCWVVVRLDG RNFHRFAEKH NFAKPNDSRA LQLMTKCAQT VMEELEDIVI AYGQSDEYSF VFKRKTNWFK RRASKFMTHV ASQFASSYVF YWRDYFEDQP LLYPPGFDGR VVVYPSNQTL KDYLSWRQAD CHINNLYNTV FWALIQQSGL TPVQAQGRLQ GTLAADKNEI LFSEFNINYN NELPMYRKGT VLIWQKVDEV MTKEIKLPTE MEGKKMAVTR TRTKPVPLHC DIIGDAFWKE HPEILDEDS.

Adds a GMP to the 5'-end of tRNA(His) after transcription and RNase P cleavage. This step is essential for proper recognition of the tRNA and for the fidelity of protein synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THG1L
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4F11
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene THG1L
Gene Accession No. Q9NWX6
Gene Alias hTHG1; ICF45; IHG-1; induced by high glucose-1; interphase cyctoplasmic foci protein 45; interphase cytoplasmic foci protein 45; Probable tRNA(His) guanylyltransferase; Thg1l; tRNA-histidine guanylyltransferase; tRNA-histidine guanylyltransferase 1 like; tRNA-histidine guanylyltransferase 1-like; tRNA-histidine guanylyltransferase 1-like (S. cerevisiae); zgc:101609
Gene Symbols THG1L
Host Species Mouse
Immunogen THG1L (AAH01523, 1 a.a. ~ 269 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54974
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.