Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiamine Nitrate 98.0+%, TCI America™
SDP

Catalog No. T0182100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0182100G 100 g
T018225G 25 g
T0182500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0182100G Supplier TCI America Supplier No. T0182100G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 532-43-4
Molecular Formula C12H17N5O4S
Molecular Weight (g/mol) 327.36
MDL Number MFCD00036330
InChI Key UIERGBJEBXXIGO-UHFFFAOYSA-N
Synonym thiamine nitrate, thiamine mononitrate, vitamin b1 nitrate, vitamin b1 mononitrate, aneurine mononitrate, aneurine nitrate, betabion mononitrate, thiamine nitrate salt, vitamin b sub 1 nitrate, thiamine mononitrate usp
PubChem CID 10762
IUPAC Name 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-3H-1λ⁴,3-thiazol-1-ylium nitrate
SMILES [O-][N+]([O-])=O.CC1=C(CCO)[S+]=CN1CC1=CN=C(C)N=C1N

Specifications

Melting Point 190°C
Color White-Yellow
pH 6.5 to 8.0 (10g/L H2O 20°C)
Quantity 100 g
Formula Weight 327.36
Percent Purity ≥98.0% (HPLC,E)
Physical Form Crystalline Powder
Chemical Name or Material Thiamine Nitrate
RTECSNumber XI7400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.