Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thianthrene 98.0+%, TCI America™
SDP

Catalog No. T018425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T018425G 25 g
T01845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T018425G Supplier TCI America Supplier No. T018425G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 92-85-3
Molecular Formula C12H8S2
Molecular Weight (g/mol) 216.32
MDL Number MFCD00005065
InChI Key GVIJJXMXTUZIOD-UHFFFAOYSA-N
PubChem CID 7109
ChEBI CHEBI:64511
IUPAC Name thianthrene
SMILES S1C2=CC=CC=C2SC2=CC=CC=C12

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 25 g
Formula Weight 216.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Thianthrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.