Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
THIK-2 Mouse anti-Mouse, Rat, Clone: N468/37, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $463.74
Specifications
| Antigen | THIK-2 | 
|---|---|
| Clone | N468/37 | 
| Concentration | 1.0 mg/mL | 
| Applications | Immunohistochemistry (Paraffin) | 
| Classification | Monoclonal | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
| MABN242325 | MilliporeSigma MABN242325UG | 25 μg | 
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $182.31
                                                 |  | |||||
| MABN2423100 | MilliporeSigma MABN2423100UG | 100 μg | 
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $463.74
                                                 |  | |||||
Description
THIK-2 Monoclonal antibody specifically detects THIK-2 in Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin).Specifications
| THIK-2 | |
| 1.0 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| IgG1 κ | |
| Protein G purified | 
| N468/37 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Mouse, Rat | |
| A recombinant fragment corresponding to 106 amino acids from the C-terminal, cytoplasmic domain of mouse THIK-2 (MNTK1). | |
| Primary | |
| Stable for 1 year at +2°C to +8°C from date of receipt. | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            .jpg-250.jpg)