Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thimet Oligopeptidase/THOP1 Antibody (4D6), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB400146W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400146W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400146W Supplier Novus Biologicals Supplier No. NB400146AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Thimet Oligopeptidase/THOP1 Monoclonal specifically detects Thimet Oligopeptidase/THOP1 in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Thimet Oligopeptidase/THOP1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4D6
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Accession No. P52888
Gene Alias EC 3.4.24, EC 3.4.24.15, Endopeptidase 24.15, EP24.15, MEPD_HUMAN, MP78, thimet oligopeptidase, thimet oligopeptidase 1, TOP
Gene Symbols THOP1
Host Species Mouse
Immunogen A synthetic peptide corresponding to residues 56-72 of human Thimet Oligopeptidase. [UniProt# P52888]
Molecular Weight of Antigen 80 kDa
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7064
Target Species Human, Mouse, Rat, Primate, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.