Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thimet Oligopeptidase/THOP1 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB053523 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB053523 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB053523 Supplier Novus Biologicals Supplier No. NBP298355DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Thimet Oligopeptidase/THOP1 Polyclonal specifically detects Thimet Oligopeptidase/THOP1 in Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Thimet Oligopeptidase/THOP1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.24, EC 3.4.24.15, Endopeptidase 24.15, EP24.15, MEPD_HUMAN, MP78, thimet oligopeptidase, thimet oligopeptidase 1, TOP
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Thimet Oligopeptidase/THOP1 (NP_073144.3; Lys 2-Cys687).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7064
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.