Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wilmad-LabGlass™ Thin Walled High Throughput NMR Tubes
GSA_VA

Catalog No. p-5211424 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Click to view available options
Length (Metric):
103.5 mm
178 mm
203 mm
Field Strength:
600 MHz
High-throughput
Quantity:
100/Pk.

Possess an average camber of 60 microns to guarantee the clarity of the spectrum for small molecule (MW<250) samples up to 600 MHz. Wilmad Thin Walled High Throughput NMR Tubes are designed for routine use in most low to mid field NMR spectrometers.

  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • Designed for routine use in most low to mid field NMR spectrometers
  • One of the best OD tolerances in the industry
  • Guaranteed to fit tightly in spinner turbine
  • Made from ASTM E438 Type 1 Class B glass
  • 100% inspected for surface defects and physical dimension to ensure the success of your experiments
  • WG-1000-4-SJ and WG-1000-7-SJ are supplied with authentic Bruker SampleJet caps, all others come with a disposable NMR cap
Note: For NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, please refer to Wilmad Precision NMR Tubes.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Length (Metric) 103.5 mm
Material ASTM E438 Type 1 Class B Glass
Camber (Metric) 60 μm
Product Type NMR Tube
Closure Type Bruker SampleJet Cap
Diameter (Metric) Outer 5 mm
Field Strength 600 MHz
Length (English) 4 in.
Thickness (Metric) Wall 0.43 mm
Quantity 100/Pk.
Model WG-1000-4-SJ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.