Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiodiglycolic acid, 98%

Catalog No. p-200150
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Plastic bottle

CAS: 123-93-3 | C4H6O4S | 150.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-93-3
Molecular Formula C4H6O4S
Molecular Weight (g/mol) 150.148
MDL Number MFCD00004359
InChI Key UVZICZIVKIMRNE-UHFFFAOYSA-N
Synonym thiodiglycolic acid, 2,2'-thiodiacetic acid, thiodiacetic acid, mercaptodiacetic acid, tdga, dicarboxymethyl sulfide, thiodi acetic acid, carboxymethylthio acetic acid, 2,2'-thiodiethanoic acid, thiodiglycollic acid
PubChem CID 31277
ChEBI CHEBI:35012
IUPAC Name 2-(carboxymethylsulfanyl)acetic acid
SMILES C(C(=O)O)SCC(=O)O

Specifications

Melting Point 128.0°C to 131.0°C
Color Gray to White
Assay Percent Range 98%
Linear Formula S(CH2CO2H)2
Beilstein 03, 253
Merck Index 15, 9484
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, insoluble in benzene
Formula Weight 150.15
Percent Purity 98%
Quantity 25 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder
Chemical Name or Material Thiodiglycolic acid, 98%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-663-9
RTECSNumber AJ6475000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.