Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiomorpholine 1,1-Dioxide 98.0+%, TCI America™
SDP

Catalog No. T219325G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 39093-93-1
Molecular Formula C4H9NO2S
Molecular Weight (g/mol) 135.181
InChI Key NDOVLWQBFFJETK-UHFFFAOYSA-N
Synonym 1,1-Dioxothiomorpholine
PubChem CID 6484228
IUPAC Name 1,4-thiazinane 1,1-dioxide
SMILES C1CS(=O)(=O)CCN1

Specifications

Melting Point 70°C
Color White
Boiling Point 155°C
Quantity 25 g
Formula Weight 135.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Thiomorpholine 1,1-Dioxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.