Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thionyl chloride, 1M soln. in dichloromethane, Thermo Scientific Chemicals

Catalog No. p-7030954
Click to view available options
Quantity:
100 mL
500 mL

Sulfuryl chloride and thionyl chloride have been found to be new chlorinating agents for alkyl sulfides. Preparation of aromatic polyesters by direct polycondensation with thionyl chloride in pyridine. Conversion of aromatic and. alpha.,. beta.-unsaturated aldehydes to dichlorides by thionyl chloride and dimethylformamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7719-09-7
MDL Number MFCD00011449

Specifications

Molecular Formula Cl3P
Linear Formula SOCl2
UN Number UN2922
Concentration 1M soln. in dichloromethane
Density 1.35
Synonym phosphorus trichloride, phosphorous trichloride, trichlorophosphine, phosphorus chloride, phosphorous chloride, phosphorus iii chloride, pcl3, phosphorus chloride pcl3, phosphine, trichloro, fosfortrichloride
Solubility Information Reacts with water.
InChI Key FAIAAWCVCHQXDN-UHFFFAOYSA-N
SMILES P(Cl)(Cl)Cl
IUPAC Name trichlorophosphane
Molecular Weight (g/mol) 137.32
PubChem CID 24387
ChEBI CHEBI:30334
Formula Weight 118.97
Quantity 100 mL
Sensitivity Moisture sensitive; May form a precipitate
Chemical Name or Material Thionyl chloride
Show More Show Less
Hazard Category H314-H335-H350
Hazard Statement GHS H Statement
H314-H351
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Precautionary Statement P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 231-748-8
RTECSNumber XM5150000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.