Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioredoxin-1 Antibody (3G7), Novus Biologicals™
SDP

Catalog No. NB298446 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298446 50 μL
NB298447 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298446 Supplier Novus Biologicals Supplier No. NBP32626550UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Thioredoxin-1 Monoclonal antibody specifically detects Thioredoxin-1 in Human, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen Thioredoxin-1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3G7
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias TRDX,TRX,TRX1,Trx80,TXN
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Thioredoxin-1 [UniProt P10599]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Breast Cancer, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7295
Target Species Human, Rat
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.