Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioredoxin Reductase 1/TRXR1 Antibody (30F1), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB308844 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308844 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308844 Supplier Novus Biologicals Supplier No. NBP259489APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Thioredoxin Reductase 1/TRXR1 Monoclonal antibody specifically detects Thioredoxin Reductase 1/TRXR1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Thioredoxin Reductase 1/TRXR1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 30F1
Conjugate APC
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias EC 1.8.1, EC 1.8.1.9, Gene associated with retinoic and IFN-induced mortality 12 protein, Gene associated with retinoic and interferon-induced mortality 12 protein, GRIM12, GRIM-12TR1, KDRF, KM-102-derived reductase-like factor, MGC9145, oxidoreductase, thioredoxin reductase 1, thioredoxin reductase 1, cytoplasmic, thioredoxin reductase GRIM-12, Thioredoxin reductase TR1, TR, Trxr1, TXNRTRXR1
Host Species Mouse
Immunogen Recombinant human Thioredoxin Reductase 1/TRXR1 (161-647aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7296
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.